Assay Details
Gene Symbol: Ext1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130540.1 | NP_001124012.1 | 6 - 7 | 1652 | 124 | |
GenBank mRNA | BC105839.1 | - | 6 - 7 | 967 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
