Assay Details
Gene Symbol: Nab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134874.1 | NP_001128346.1 | 2 - 3 | 1163 | 56 | |
XM_006241459.2 | XP_006241521.1 | 2 - 3 | 2671 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
