Assay Details
Gene Symbol: Nab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134874.1 | NP_001128346.1 | 4 - 5 | 1354 | 65 | |
XM_006241459.2 | XP_006241521.1 | 4 - 5 | 2862 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
