Assay Details
Gene Symbol: Gtse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130500.1 | NP_001123972.1 | 1 - 2 | 123 | 129 | |
XM_006242160.2 | XP_006242222.1 | 2 - 3 | 227 | 129 | ||
GenBank mRNA | FQ218328.1 | - | 2 - 3 | 195 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
