Assay Details
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139080.2 | NP_620780.2 | 3 - 4 | 268 | 75 | |
XM_008765671.1 | XP_008763893.1 | 3 - 4 | 261 | 75 | ||
GenBank mRNA | AB066102.1 | - | 2 - 3 | 252 | 75 | |
BC078718.1 | - | 3 - 4 | 268 | 7115290 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
