Assay Details
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139080.2 | NP_620780.2 | 5 - 6 | 847 | 67 | |
XM_008765671.1 | XP_008763893.1 | 5 - 6 | 840 | 67 | ||
GenBank mRNA | AB066102.1 | - | 4 - 5 | 831 | 67 | |
BC078718.1 | - | 5 - 6 | 847 | 7115290 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
