Assay Details
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139080.2 | NP_620780.2 | 6 - 7 | 1038 | 143 | |
XM_008765671.1 | XP_008763893.1 | 6 - 7 | 1031 | 143 | ||
GenBank mRNA | AB066102.1 | - | 5 - 6 | 1022 | 143 | |
BC078718.1 | - | 6 - 7 | 1038 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
