Assay Details
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139080.2 | NP_620780.2 | 9 - 10 | 1504 | 89 | |
XM_008765671.1 | XP_008763893.1 | 8 - 9 | 1335 | 89 | ||
GenBank mRNA | AB066102.1 | - | 8 - 9 | 1488 | 89 | |
BC078718.1 | - | 9 - 10 | 1504 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
