Assay Details
Gene Symbol: Ndrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011991.1 | NP_001011991.1 | 13 - 14 | 990 | 96 | |
XM_006241708.2 | XP_006241770.1 | 13 - 14 | 1262 | 96 | ||
GenBank mRNA | AY500369.1 | - | 13 - 14 | 952 | 96 | |
BC081898.1 | - | 13 - 14 | 990 | 7110455 | 96 | |
FQ212154.1 | - | 13 - 14 | 1002 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
