Assay Details
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013104.1 | NP_001013122.1 | 2 - 3 | 1224 | 73 | |
XM_008765549.1 | XP_008763771.1 | 2 - 3 | 3288 | 73 | ||
XM_008765550.1 | XP_008763772.1 | 2 - 3 | 1349 | 73 | ||
XM_008765551.1 | XP_008763773.1 | 4 - 5 | 1341 | 73 | ||
XM_008765552.1 | XP_008763774.1 | 4 - 5 | 1322 | 73 | ||
XM_008765553.1 | XP_008763775.1 | 2 - 3 | 1241 | 73 | ||
XM_008765554.1 | XP_008763776.1 | 2 - 3 | 1352 | 73 | ||
XM_008765557.1 | XP_008763779.1 | 2 - 3 | 228 | 73 | ||
XM_008765558.1 | XP_008763780.1 | 2 - 3 | 235 | 73 | ||
XM_008765559.1 | XP_008763781.1 | 2 - 3 | 215 | 73 | ||
GenBank mRNA | BC079391.1 | - | 2 - 3 | 1224 | 73 | |
FQ220813.1 | - | 2 - 3 | 185 | 73 | ||
FQ225933.1 | - | 2 - 3 | 220 | 73 | ||
FQ227443.1 | - | 2 - 3 | 196 | 73 | ||
FQ233760.1 | - | 2 - 3 | 176 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
