Assay Details
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013104.1 | NP_001013122.1 | 5 - 6 | 1497 | 64 | |
XM_008765549.1 | XP_008763771.1 | 6 - 7 | 3618 | 64 | ||
XM_008765550.1 | XP_008763772.1 | 6 - 7 | 1679 | 64 | ||
XM_008765551.1 | XP_008763773.1 | 8 - 9 | 1671 | 64 | ||
XM_008765552.1 | XP_008763774.1 | 8 - 9 | 1652 | 64 | ||
XM_008765553.1 | XP_008763775.1 | 6 - 7 | 1571 | 64 | ||
XM_008765554.1 | XP_008763776.1 | 5 - 6 | 1625 | 64 | ||
XM_008765555.1 | XP_008763777.1 | 5 - 6 | 1610 | 64 | ||
XM_008765556.1 | XP_008763778.1 | 4 - 5 | 1553 | 64 | ||
XM_008765557.1 | XP_008763779.1 | 5 - 6 | 501 | 64 | ||
XM_008765558.1 | XP_008763780.1 | 5 - 6 | 508 | 64 | ||
XM_008765559.1 | XP_008763781.1 | 5 - 6 | 488 | 64 | ||
XM_008765560.1 | XP_008763782.1 | 5 - 6 | 486 | 64 | ||
XM_008765561.1 | XP_008763783.1 | 4 - 5 | 429 | 64 | ||
XM_008765562.1 | XP_008763784.1 | 4 - 5 | 414 | 64 | ||
GenBank mRNA | BC079391.1 | - | 5 - 6 | 1497 | 64 | |
FQ225933.1 | - | 5 - 6 | 493 | 64 | ||
FQ227443.1 | - | 5 - 6 | 469 | 64 | ||
FQ233760.1 | - | 5 - 6 | 449 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
