Assay Details
Gene Symbol: Stk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031735.1 | NP_113923.1 | 5 - 6 | 605 | 114 | |
XM_008765458.1 | XP_008763680.1 | 5 - 6 | 783 | 114 | ||
XM_008765459.1 | XP_008763681.1 | 5 - 6 | 783 | 114 | ||
XM_008765460.1 | XP_008763682.1 | 5 - 6 | 472 | 114 | ||
GenBank mRNA | AJ001529.1 | - | 5 - 6 | 605 | 114 | |
BC161844.1 | - | 5 - 6 | 688 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
