Assay Details
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012121.1 | NP_001012121.1 | 2 - 3 | 397 | 80 | |
XM_008765722.1 | XP_008763944.1 | 3 - 4 | 1493 | 80 | ||
XM_008765723.1 | XP_008763945.1 | 2 - 3 | 253 | 80 | ||
XM_008765724.1 | XP_008763946.1 | 2 - 3 | 1078 | 80 | ||
XM_008765725.1 | XP_008763947.1 | 3 - 4 | 230 | 80 | ||
XM_008765726.1 | XP_008763948.1 | 3 - 4 | 225 | 80 | ||
GenBank mRNA | BC090007.1 | - | 2 - 3 | 397 | 7323849 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008765723.1, XM_008765724.1, XM_008765725.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
