Assay Details
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012121.1 | NP_001012121.1 | 4 - 5 | 498 | 63 | |
XM_008765722.1 | XP_008763944.1 | 5 - 6 | 1594 | 63 | ||
XM_008765723.1 | XP_008763945.1 | 4 - 5 | 354 | 63 | ||
XM_008765724.1 | XP_008763946.1 | 4 - 5 | 1179 | 63 | ||
XM_008765725.1 | XP_008763947.1 | 5 - 6 | 331 | 63 | ||
XM_008765726.1 | XP_008763948.1 | 5 - 6 | 326 | 63 | ||
GenBank mRNA | BC090007.1 | - | 4 - 5 | 498 | 7323849 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008765723.1, XM_008765724.1, XM_008765725.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
