Assay Details
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012121.1 | NP_001012121.1 | 6 - 7 | 735 | 84 | |
XM_008765722.1 | XP_008763944.1 | 7 - 8 | 1831 | 84 | ||
XM_008765723.1 | XP_008763945.1 | 6 - 7 | 591 | 84 | ||
XM_008765724.1 | XP_008763946.1 | 6 - 7 | 1416 | 84 | ||
XM_008765725.1 | XP_008763947.1 | 7 - 8 | 568 | 84 | ||
XM_008765726.1 | XP_008763948.1 | 7 - 8 | 563 | 84 | ||
GenBank mRNA | BC090007.1 | - | 6 - 7 | 735 | 7323849 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
