Assay Details
Gene Symbol: Cpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765417.1 | XP_008763639.1 | 1 - 2 | 198 | 81 | |
GenBank mRNA | BC070884.1 | - | 1 - 2 | 58 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC070884.1, XM_008765417.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
