Assay Details
Gene Symbol: Cpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031640.1 | NP_113828.1 | 3 - 4 | 551 | 60 | |
XM_008765417.1 | XP_008763639.1 | 2 - 3 | 652 | 60 | ||
GenBank mRNA | AF097723.1 | - | 3 - 4 | 551 | 60 | |
AF131077.1 | - | 2 - 3 | 493 | 60 | ||
BC070884.1 | - | 2 - 3 | 512 | 60 | ||
FQ210652.1 | - | 1 - 2 | 291 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
