Assay Details
Gene Symbol: Cpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031640.1 | NP_113828.1 | 6 - 7 | 1081 | 59 | |
XM_008765417.1 | XP_008763639.1 | 5 - 6 | 1182 | 59 | ||
GenBank mRNA | AF097723.1 | - | 6 - 7 | 1081 | 59 | |
AF131077.1 | - | 5 - 6 | 1023 | 59 | ||
BC070884.1 | - | 5 - 6 | 1042 | 59 | ||
FQ210652.1 | - | 4 - 5 | 821 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
