Assay Details
Gene Symbol: Emc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113785.1 | NP_001107257.1 | 1 - 2 | 61 | 145 | |
GenBank mRNA | BC158805.1 | - | 1 - 2 | 61 | 7322060 | 145 |
FQ215024.1 | - | 1 - 2 | 50 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
