Assay Details
Gene Symbol: Emc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113785.1 | NP_001107257.1 | 5 - 6 | 469 | 64 | |
GenBank mRNA | BC158805.1 | - | 5 - 6 | 469 | 7322060 | 64 |
FQ214096.1 | - | 2 - 3 | 352 | 64 | ||
FQ215024.1 | - | 5 - 6 | 458 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
