Assay Details
Gene Symbol: Smpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006997.1 | NP_001006998.1 | 3 - 4 | 1336 | 72 | |
GenBank mRNA | BC083780.1 | - | 3 - 4 | 1336 | 7096039 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
