Assay Details
Gene Symbol: Mrpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106282.1 | NP_001099752.1 | 6 - 7 | 551 | 114 | |
GenBank mRNA | FQ212514.1 | - | 6 - 7 | 748 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
