Assay Details
Gene Symbol: Mrpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106282.1 | NP_001099752.1 | 7 - 8 | 644 | 127 | |
GenBank mRNA | FQ212514.1 | - | 7 - 8 | 841 | 127 | |
FQ214311.1 | - | 7 - 8 | 693 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52339 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14847 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14847 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14847 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14847 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92379 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10723986 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723984 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723988 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723987 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723983 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723985 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10723982 | - | - |
Rat Genome 230 2.0 Array | 1371586_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379306_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379306_at | - | - |
Rat Genome 230 2.0 Array | 1371586_at | - | - |
Affymetrix HT Rat Focus Array | 1379306_at | - | - |
Affymetrix HT Rat Focus Array | 1371586_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1371586_at | - | - |
Affymetrix HT Rat Focus Array | 1379306_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100048750.rn.1 | TC0100006142.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100048746.rn.1 | TC0100006142.rn.1 | RTA TC |
Rat Expression Set 230 | 1371586_at | - | - |
Rat Expression Set 230 | 1371586_PM_at | - | - |
Rat Expression Set 230 | 1379306_PM_at | - | - |
Rat Expression Set 230 | 1379306_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723982 | - | - |
Rat Genome U34 Set | rc_AI014012_at | - | - |
Rat Genome U34 Set | rc_AI103834_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21663980 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353002 | - | - |
