Assay Details
Gene Symbol: Ric3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115045.3 | NP_001108517.1 | 1 - 2 | 126 | 94 | |
NM_001277069.1 | NP_001263998.1 | 1 - 2 | 126 | 94 | ||
GenBank mRNA | AM422212.1 | - | 1 - 2 | 126 | 94 | |
AM422213.1 | - | 1 - 2 | 126 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
