Assay Details
Gene Symbol: Ric3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115045.3 | NP_001108517.1 | 4 - 5 | 519 | 100 | |
GenBank mRNA | AM422213.1 | - | 4 - 5 | 519 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
