Assay Details
Gene Symbol: Dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012345.1 | NP_001012345.1 | 5 - 6 | 816 | 86 | |
XM_006229733.2 | XP_006229795.1 | 4 - 5 | 856 | 86 | ||
XM_008759689.1 | XP_008757911.1 | 5 - 6 | 567 | 86 | ||
GenBank mRNA | AJ487787.1 | - | 1 - 2 | 112 | 86 | |
BC089846.1 | - | 5 - 6 | 816 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
