Assay Details
Gene Symbol: Far1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011933.1 | NP_001011933.1 | 10 - 11 | 1312 | 147 | |
XM_006230017.2 | XP_006230079.1 | 11 - 12 | 1394 | 147 | ||
XM_006230018.2 | XP_006230080.1 | 11 - 12 | 1319 | 147 | ||
XM_006230020.2 | XP_006230082.1 | 11 - 12 | 1396 | 147 | ||
GenBank mRNA | BC082015.1 | - | 10 - 11 | 1312 | 147 | |
FQ221385.1 | - | 11 - 12 | 1378 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
