Assay Details
Gene Symbol: Far1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011933.1 | NP_001011933.1 | 3 - 4 | 425 | 85 | |
XM_006230017.2 | XP_006230079.1 | 4 - 5 | 507 | 85 | ||
XM_006230018.2 | XP_006230080.1 | 4 - 5 | 432 | 85 | ||
XM_006230020.2 | XP_006230082.1 | 4 - 5 | 509 | 85 | ||
GenBank mRNA | BC082015.1 | - | 3 - 4 | 425 | 7132096 | 85 |
FQ221385.1 | - | 4 - 5 | 491 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
