Assay Details
Gene Symbol: Far1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011933.1 | NP_001011933.1 | 5 - 6 | 781 | 108 | |
XM_006230017.2 | XP_006230079.1 | 6 - 7 | 863 | 108 | ||
XM_006230018.2 | XP_006230080.1 | 6 - 7 | 788 | 108 | ||
XM_006230020.2 | XP_006230082.1 | 6 - 7 | 865 | 108 | ||
GenBank mRNA | BC082015.1 | - | 5 - 6 | 781 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
