Assay Details
Gene Symbol: Ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013167.2 | NP_037299.1 | 3 - 4 | 458 | 93 | |
GenBank mRNA | AB006614.1 | - | 2 - 3 | 330 | 93 | |
AF030163.1 | - | 2 - 3 | 345 | 93 | ||
AF035943.1 | - | 2 - 3 | 341 | 93 | ||
BC072546.1 | - | 3 - 4 | 458 | 7105743 | 93 | |
U92069.1 | - | 3 - 4 | 465 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
