Assay Details
Gene Symbol: Ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013167.2 | NP_037299.1 | 1 - 2 | 92 | 123 | |
GenBank mRNA | BC072546.1 | - | 1 - 2 | 92 | 7105743 | 123 |
U92069.1 | - | 1 - 2 | 99 | 123 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC072546.1,NM_013167.2,U92069.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
