Assay Details
Gene Symbol: Il4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133380.2 | NP_596871.2 | 7 - 8 | 845 | 82 | |
XM_006230215.2 | XP_006230277.1 | 8 - 9 | 996 | 82 | ||
XM_006230216.2 | XP_006230278.1 | 6 - 7 | 786 | 82 | ||
XM_008759862.1 | XP_008758084.1 | 7 - 8 | 6204 | 82 | ||
XM_008759863.1 | XP_008758085.1 | 8 - 9 | 6151 | 82 | ||
XM_008759864.1 | XP_008758086.1 | 8 - 9 | 6151 | 82 | ||
GenBank mRNA | AB015747.1 | - | 7 - 8 | 852 | 82 | |
X69903.1 | - | 683 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
