Assay Details
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271536.1 | NP_001258465.1 | 1 - 2 | 128 | 98 | |
XM_006230211.1 | XP_006230273.1 | 2 - 3 | 155 | 98 | ||
GenBank mRNA | CB750468.1 | - | 1 - 2 | 128 | 98 | |
FQ228954.1 | - | 1 - 2 | 74 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271536.1, XM_006230211.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
