Assay Details
Gene Symbol: Il18bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053374.2 | NP_445826.2 | 3 - 4 | 412 | 64 | |
XM_008759805.1 | XP_008758027.1 | 3 - 4 | 2570 | 64 | ||
GenBank mRNA | AF154569.1 | - | 2 - 3 | 223 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
