Assay Details
Gene Symbol: Arl6ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198737.2 | NP_942032.1 | 1 - 2 | 90 | 117 | |
GenBank mRNA | AY316590.1 | - | 1 - 2 | 91 | 117 | |
BC099071.1 | - | 1 - 2 | 90 | 117 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY316590.1, BC099071.1, NM_198737.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
