Assay Details
Gene Symbol: Arl6ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198737.2 | NP_942032.1 | 3 - 4 | 352 | 109 | |
GenBank mRNA | AY316590.1 | - | 3 - 4 | 353 | 109 | |
BC099071.1 | - | 3 - 4 | 352 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
