Assay Details
Gene Symbol: Doc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022937.2 | NP_075226.1 | 5 - 6 | 719 | 115 | |
XM_006230335.2 | XP_006230397.1 | 6 - 7 | 819 | 115 | ||
XM_006230336.1 | XP_006230398.1 | 6 - 7 | 1003 | 115 | ||
XM_006230337.2 | XP_006230399.1 | 6 - 7 | 815 | 115 | ||
XM_006230339.2 | XP_006230401.1 | 6 - 7 | 753 | 115 | ||
XM_006230340.1 | XP_006230402.1 | 6 - 7 | 937 | 115 | ||
XM_006230341.2 | XP_006230403.1 | 6 - 7 | 937 | 115 | ||
XM_008759918.1 | XP_008758140.1 | 6 - 7 | 850 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
