Assay Details
Gene Symbol: Rab6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053366.1 | NP_445818.1 | 6 - 7 | 938 | 133 | |
XM_006229794.1 | XP_006229856.1 | 6 - 7 | 1023 | 133 | ||
GenBank mRNA | AF148210.1 | - | 3 - 4 | 291 | 133 | |
FQ212069.1 | - | 6 - 7 | 965 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
