Assay Details
Gene Symbol: Rab6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229794.1 | XP_006229856.1 | 4 - 5 | 818 | 119 | |
GenBank mRNA | AF148210.1 | - | 1 - 2 | 86 | 119 | |
FQ212069.1 | - | 4 - 5 | 760 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
