Assay Details
Gene Symbol: Ppp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230206.1 | XP_006230268.1 | 1 - 2 | 111 | 118 | |
GenBank mRNA | FQ216602.1 | - | 1 - 2 | 71 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006230206.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
