Assay Details
Gene Symbol: Ppp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134359.1 | NP_599186.1 | 1 - 2 | 279 | 86 | |
XM_006230206.1 | XP_006230268.1 | 2 - 3 | 275 | 86 | ||
GenBank mRNA | BC091574.1 | - | 1 - 2 | 279 | 7135215 | 86 |
FQ216602.1 | - | 2 - 3 | 235 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
