Assay Details
Gene Symbol: Mylpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012605.2 | NP_036737.1 | 3 - 4 | 216 | 65 | |
GenBank mRNA | BI278596.1 | - | 4 - 5 | 65 | ||
CB770854.1 | - | 3 - 4 | 226 | 65 | ||
FQ215085.1 | - | 3 - 4 | 249 | 65 | ||
FQ216891.1 | - | 3 - 4 | 232 | 65 | ||
FQ216934.1 | - | 3 - 4 | 232 | 65 | ||
FQ217098.1 | - | 3 - 4 | 232 | 65 | ||
FQ217115.1 | - | 1 - 2 | 81 | 65 | ||
FQ217155.1 | - | 3 - 4 | 232 | 65 | ||
FQ217219.1 | - | 3 - 4 | 232 | 65 | ||
FQ217259.1 | - | 3 - 4 | 232 | 65 | ||
FQ217349.1 | - | 2 - 3 | 156 | 65 | ||
FQ217365.1 | - | 3 - 4 | 231 | 65 | ||
FQ217368.1 | - | 3 - 4 | 248 | 65 | ||
FQ217533.1 | - | 3 - 4 | 260 | 65 | ||
FQ217703.1 | - | 3 - 4 | 232 | 65 | ||
FQ217740.1 | - | 3 - 4 | 248 | 65 | ||
FQ217881.1 | - | 3 - 4 | 229 | 65 | ||
FQ218017.1 | - | 3 - 4 | 232 | 65 | ||
FQ223762.1 | - | 3 - 4 | 213 | 65 | ||
FQ223868.1 | - | 3 - 4 | 232 | 65 | ||
FQ223911.1 | - | 3 - 4 | 248 | 65 | ||
FQ223944.1 | - | 2 - 3 | 98 | 65 | ||
FQ223966.1 | - | 3 - 4 | 232 | 65 | ||
FQ223985.1 | - | 3 - 4 | 232 | 65 | ||
FQ224150.1 | - | 1 - 2 | 75 | 65 | ||
FQ224231.1 | - | 3 - 4 | 232 | 65 | ||
FQ224253.1 | - | 3 - 4 | 231 | 65 | ||
FQ224391.1 | - | 3 - 4 | 230 | 65 | ||
FQ224404.1 | - | 3 - 4 | 230 | 65 | ||
FQ224511.1 | - | 3 - 4 | 232 | 65 | ||
FQ224525.1 | - | 3 - 4 | 251 | 65 | ||
FQ224720.1 | - | 3 - 4 | 232 | 65 | ||
FQ224781.1 | - | 3 - 4 | 248 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
