Assay Details
Gene Symbol: Kif22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009645.1 | NP_001009645.1 | 1 - 2 | 85 | 67 | |
GenBank mRNA | BC088421.1 | - | 1 - 2 | 85 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
