Assay Details
Gene Symbol: Ino80e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013900.1 | NP_001013922.1 | 5 - 6 | 513 | 91 | |
XM_006230243.1 | XP_006230305.1 | 5 - 6 | 436 | 91 | ||
GenBank mRNA | BC079045.1 | - | 5 - 6 | 513 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
