Assay Details
Gene Symbol: P4ha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759761.1 | XP_008757983.1 | 2 - 3 | 372 | 80 | |
XM_008759762.1 | XP_008757984.1 | 2 - 3 | 372 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
