Assay Details
Gene Symbol: P4ha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759761.1 | XP_008757983.1 | 3 - 4 | 595 | 68 | |
XM_008759762.1 | XP_008757984.1 | 3 - 4 | 595 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
