Assay Details
Gene Symbol: Folr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229842.2 | XP_006229904.1 | 4 - 5 | 635 | 55 | |
GenBank mRNA | BC157811.1 | - | 3 - 4 | 497 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
