Assay Details
Gene Symbol: Gab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053417.1 | NP_445869.1 | 4 - 5 | 1201 | 68 | |
XM_008759806.1 | XP_008758028.1 | 3 - 4 | 2833 | 68 | ||
GenBank mRNA | AF230367.2 | - | 4 - 5 | 1201 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
