Assay Details
Gene Symbol: Gab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053417.1 | NP_445869.1 | 7 - 8 | 1626 | 67 | |
XM_008759806.1 | XP_008758028.1 | 6 - 7 | 3258 | 67 | ||
GenBank mRNA | AF230367.2 | - | 7 - 8 | 1626 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
