Assay Details
Gene Symbol: Gab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053417.1 | NP_445869.1 | 9 - 10 | 1857 | 121 | |
XM_008759806.1 | XP_008758028.1 | 8 - 9 | 3489 | 121 | ||
GenBank mRNA | AF230367.2 | - | 9 - 10 | 1857 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transmembrane receptor protein tyrosine kinase signaling pathway
integrin-mediated signaling pathway
positive regulation of cell population proliferation
cell migration
osteoclast differentiation
intracellular signal transduction
positive regulation of mast cell degranulation
phosphatidylinositol-mediated signaling
transmembrane receptor protein tyrosine kinase adaptor activity
phosphatidylinositol-3,4,5-trisphosphate binding
protein domain specific binding
phosphatidylinositol-3,4-bisphosphate binding
transmembrane receptor protein tyrosine kinase signaling pathway
integrin-mediated signaling pathway
positive regulation of cell population proliferation
cell migration
osteoclast differentiation
intracellular signal transduction
positive regulation of mast cell degranulation
phosphatidylinositol-mediated signaling
transmembrane receptor protein tyrosine kinase adaptor activity
phosphatidylinositol-3,4,5-trisphosphate binding
protein domain specific binding
phosphatidylinositol-3,4-bisphosphate binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P144964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121292 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P144964 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P144964 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P144964 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P529077 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95408 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70597 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84337 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61736 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85685 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99560 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98969 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37622 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59473 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82610 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22478 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10708750 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708749 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708747 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708755 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708748 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708753 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708746 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708751 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708754 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708752 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10708745 | - | - |
Rat Genome 230 2.0 Array | 1369458_at | - | - |
Rat Genome 230 2.0 Array | 1369458_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369458_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369458_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100015879.rn.1 | TC0100002070.rn.1 | RTA TC |
Rat Expression Set 230 | 1369458_PM_at | - | - |
Rat Expression Set 230 | 1369458_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708745 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21665263 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353793 | - | - |
RatRef-12 Expression BeadChip | ILMN_61395 | - | - |
